Learn More
Gold Biotechnology Inc GV3101 pSoup P19 Agro Electro
SDP
Catalog No. 5000562283
Encompass_Preferred
GoldBio s GV3101 electrocompetent Agrobacterium cells are a highly efficient tool for genetic transformation of dicotyledonous and monocotyledonous plant species such as Arabidopsis thaliana tobacco potato and corn These cells carry the nopaline-type Ti plasmid pMP90 (pTiC58DT-DNA) that contains the vir genes required for the transfer and integration of T-DNA into the plant genome Additionally the cells are resistant to rifampicin gentamicin and tetracycline which allows for selection of transformed cells
Catalog No. 50-005-62283 Supplier Gold Biotechnology Inc Supplier No. CC22715X50
GoldBio s GV3101 electrocompetent Agrobacterium cells are a highly efficient tool for genetic transformation of dicotyledonous and monocotyledonous plant species such as Arabidopsis thaliana tobacco potato and corn These cells carry the nopaline-type Ti plasmid pMP90 (pTiC58DT-DNA) that contains the vir genes required for the transfer and integration of T-DNA into the plant genome Additionally the cells are resistant to rifampicin gentamicin and tetracycline which allows for selection of transformed cells