Learn More
Gold Biotechnology Inc GV3101 pSuperAgro v4tet Electr
SDP
Catalog No. 5000562307
Encompass_Preferred
GoldBio s GV3101 Agrobacterium Electrocompetent Cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our GV3101 strain harbors the C58 chromosomal backbone containing rifampicin resistance and the Ti plasmid pMP90 (pTiC58DT-DNA) harboring the gentamicin resistance A functional T-DNA binary system can be built using our GV3101 strains as the T-DNA region has been deleted from the Ti plasmid and instead has a binary vector containing the missing T-region The binary system makes possible to transfer genetic material into a host plant s genome Our system is often used for Agrobacterium -mediated transformation in mono and dicotyledonous species such as Arabidopsis thaliana tobacco potato soybeans and corn
Catalog No. 50-005-62307 Supplier Gold Biotechnology Inc Supplier No. CC6145X50
GoldBio s GV3101 Agrobacterium Electrocompetent Cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction Our GV3101 strain harbors the C58 chromosomal backbone containing rifampicin resistance and the Ti plasmid pMP90 (pTiC58DT-DNA) harboring the gentamicin resistance A functional T-DNA binary system can be built using our GV3101 strains as the T-DNA region has been deleted from the Ti plasmid and instead has a binary vector containing the missing T-region The binary system makes possible to transfer genetic material into a host plant s genome Our system is often used for Agrobacterium -mediated transformation in mono and dicotyledonous species such as Arabidopsis thaliana tobacco potato soybeans and corn