Learn More
Complement Technology Inc GVBE (GVBo with EDTA) 250ml
Catalog No. NC1528192
Encompass
GVBE is an isotonic buffer used in complement assays to prevent complement activation or to stop ongoing complement activation when added at the end of an incubation
Catalog No. NC1528192 Supplier Complement Technology Inc Supplier No. B104
GVBE is an isotonic buffer used in complement assays to prevent complement activation or to stop ongoing complement activation when added at the end of an incubation