Learn More
Waters Corp Ferrule, Graphite/Vespel, 1/16 in. x 0.05 in. Bore
Catalog No. 50791742 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a ferrule designed for use in HPLC systems. It is constructed from Graphite/Vespel and features a 1/16 in. x 0.05 in. bore, making it suitable for secure connections within chromatographic setups.
  • Constructed from Graphite/Vespel material.
  • Designed with a 1/16 in. x 0.05 in. bore.
  • Essential component for high-performance liquid chromatography systems.
  • Categorized under tubing, ferrules, and accessories.
  • Compatible with Platform LC systems.
Catalog No. 50-791-742 Supplier Waters Corp Supplier No. 405000154
This product is a ferrule designed for use in HPLC systems. It is constructed from Graphite/Vespel and features a 1/16 in. x 0.05 in. bore, making it suitable for secure connections within chromatographic setups.
  • Constructed from Graphite/Vespel material.
  • Designed with a 1/16 in. x 0.05 in. bore.
  • Essential component for high-performance liquid chromatography systems.
  • Categorized under tubing, ferrules, and accessories.
  • Compatible with Platform LC systems.