Learn More
Trajan Scientific 1/4" GC ferrule 6 mm ID 60% Vespel / 40% graphite PK10
SDP
Catalog No. 502724148
Encompass_Preferred
1/4" GC ferrule 6 mm ID 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4148 Supplier Trajan Scientific Supplier No. 213406
1/4" GC ferrule 6 mm ID 60% Vespel / 40% graphite PK10