Learn More
Trajan Scientific 1/8" GC ferrule 0.4 mm ID 60% Vespel / 40% graphite PK10
SDP
Catalog No. 502724159
Encompass_Preferred
1/8" GC ferrule 0.4 mm ID 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4159 Supplier Trajan Scientific Supplier No. 213204
1/8" GC ferrule 0.4 mm ID 60% Vespel / 40% graphite PK10