Learn More
Trajan Scientific 1/8" GC ferrule 0.8 mm ID 85% Vespel / 15% graphite PK10
SDP
Catalog No. 502724096
Encompass_Preferred
1/8" GC ferrule 0.8 mm ID 85% Vespel / 15% graphite PK10
Catalog No. 50-272-4096 Supplier Trajan Scientific Supplier No. 215208
1/8" GC ferrule 0.8 mm ID 85% Vespel / 15% graphite PK10