Learn More
Trajan Scientific 1/8" GC ferrule 0.8 mm ID two-hole 60% Vespel / 40% graphite PK10
SDP
Catalog No. 502724154
Encompass_Preferred
1/8" GC ferrule 0.8 mm ID two-hole 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4154 Supplier Trajan Scientific Supplier No. 213228
1/8" GC ferrule 0.8 mm ID two-hole 60% Vespel / 40% graphite PK10