Learn More
Trajan Scientific 1/8" GC ferrule 1/8" ID 60% Vespel / 40% graphite PK10
SDP
Catalog No. 502724160
Encompass_Preferred
1/8" GC ferrule 1/8" ID 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4160 Supplier Trajan Scientific Supplier No. 213200
1/8" GC ferrule 1/8" ID 60% Vespel / 40% graphite PK10