Learn More
Trajan Scientific 1/16" GC ferrule 0.8 mm ID (long) 85% Vespel / 15% graphite PK10
SDP
Catalog No. 502724115
Encompass_Preferred
1/16" GC ferrule 0.8 mm ID (long) 85% Vespel / 15% graphite PK10
Catalog No. 50-272-4115 Supplier Trajan Scientific Supplier No. 215108
1/16" GC ferrule 0.8 mm ID (long) 85% Vespel / 15% graphite PK10