Learn More
Trajan Scientific 1/16" GC ferrule 0.3 and 0.5 mm ID (long) 85% Vespel / 15% Graphite PK10
SDP
Catalog No. 502724108
Encompass_Preferred
1/16" GC ferrule 0.3 and 0.5 mm ID (long) 85% Vespel / 15% Graphite PK10
Catalog No. 50-272-4108 Supplier Trajan Scientific Supplier No. 215135
1/16" GC ferrule 0.3 and 0.5 mm ID (long) 85% Vespel / 15% Graphite PK10