Learn More
Trajan Scientific 1/16" GC ferrule 0.5 mm ID (long) two-hole 60% Vespel / 40% graphite PK10
SDP
Catalog No. 502724166
Encompass_Preferred
1/16" GC ferrule 0.5 mm ID (long) two-hole 60% Vespel / 40% graphite PK10
Catalog No. 50-272-4166 Supplier Trajan Scientific Supplier No. 213125
1/16" GC ferrule 0.5 mm ID (long) two-hole 60% Vespel / 40% graphite PK10