Learn More
3M™ GVP-Series Belt-Mounted PAPRs Compatible Breathing Tubes
Catalog No. 19088270 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
GVP system with H series hood
Catalog No. For Use With (Equipment)
19-088-270 GVP system with H series hood
1 options
Catalog No. 19-088-270 Supplier 3M™ Supplier No. 7000051925
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Designed for use with 3M™ GVP Systems.
  • For use with H Series Hoods, or GVP Systems that have 3M™ Full Facepieces 7800S and 6000 DIN Series.
  • 3M Catalog# GVP-122
TRUSTED_SUSTAINABILITY
Specifications
Material Vinyl
Product Type Breathing Tube
For Use With (Application) Assembly, machine operation
For Use With (Equipment) GVP system with H series hood
Includes Breathing tube, metal thumbscrew clamp and squeeze clamp
Model GVP-122