Learn More
Medchemexpress LLC HY-50691 5mg Medchemexpress, GW-1100 CAS:306974-70-9 Purity:>98%
SDP
Catalog No. 502030947
Encompass_Preferred
Medchemexpress, HY-50691 5mg GW-1100 CAS:306974-70-9 GW-1100 is a selective GPR40 antagonist with a pIC50 of 6.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0947 Supplier Medchemexpress LLC Supplier No. HY506915MG
Medchemexpress, HY-50691 5mg GW-1100 CAS:306974-70-9 GW-1100 is a selective GPR40 antagonist with a pIC50 of 6.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.