Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GW 2580 98.0+%, TCI America™
SDP

Catalog No. G050450MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G050450MG 50 mg
G0504250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G050450MG Supplier TCI America Supplier No. G050450MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 870483-87-7
Molecular Formula C20H22N4O3
Molecular Weight (g/mol) 366.42
MDL Number MFCD12024701
InChI Key MYQAUKPBNJWPIE-UHFFFAOYSA-N
Synonym 5-[3-Methoxy-4-[(4-methoxybenzyl)oxy]benzyl]pyrimidine-2,4-diamine
PubChem CID 11617559
IUPAC Name 5-({3-methoxy-4-[(4-methoxyphenyl)methoxy]phenyl}methyl)pyrimidine-2,4-diamine
SMILES COC1=CC=C(COC2=C(OC)C=C(CC3=CN=C(N)N=C3N)C=C2)C=C1

Specifications

Quantity 50 mg
Formula Weight 366.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GW 2580
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.