Learn More
Abcam GW-311616A, 5MG
Catalog No. 502671203
Encompass_Preferred
MW 434 Da, Purity >=98%. A potent and selective intracellular neutrophil elastase (NE, alpha-1-proteinase) inhibitor. Inhibits human neutrophil elastase (HNE) and is selective over other human serine proteases with IC₅₀ values of 22 nM for HNE, >100 μM for trypsin, cathepsin G and plasmin, >3 μM for chymotrypsin and tissue plasminogen activator.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1203 Supplier Abcam Supplier No. AB2870905MG
MW 434 Da, Purity >=98%. A potent and selective intracellular neutrophil elastase (NE, alpha-1-proteinase) inhibitor. Inhibits human neutrophil elastase (HNE) and is selective over other human serine proteases with IC₅₀ values of 22 nM for HNE, >100 μM for trypsin, cathepsin G and plasmin, >3 μM for chymotrypsin and tissue plasminogen activator.
The product is subject to the following: Abcam Restricted Use Statement