Learn More
Cayman Chemical GW 3965 (hydrochloride), Purity: ≥98%, CAS: 405911-17-3, 50 mg
SDP
Catalog No. 502891252
Encompass_Preferred
An orally-active agonist of LXRα and LXRβ, activating the human isoforms with EC50 values of 190 and 30 nM, respectively; alters LXR-regulated gene expression in mice and rats, affecting pathways related to glucose and lipid metabolism
Catalog No. 50-289-1252 Supplier Cayman Chemical Supplier No. 1005450 MG
An orally-active agonist of LXRα and LXRβ, activating the human isoforms with EC50 values of 190 and 30 nM, respectively; alters LXR-regulated gene expression in mice and rats, affecting pathways related to glucose and lipid metabolism