Learn More
TARGETMOL CHEMICALS INC GW 441756 25MG
SDP
Catalog No. 502579613
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW 441756 is a specific Tropomyosin-related kinase A (TrkA) inhibitor with an IC50 value of 2 nM. purity: 99%
Catalog No. 50-257-9613 Supplier TARGETMOL CHEMICALS INC Supplier No. T6052
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW 441756 is a specific Tropomyosin-related kinase A (TrkA) inhibitor with an IC50 value of 2 nM. purity: 99%