Learn More
Enzo Life Sciences GW-441756 (50mg). CAS: 504433-23-3
SDP
Supplier:  Enzo Life Sciences BMLEI3640050
Encompass_Preferred
Potent and selective inhibitor of NGFR kinase (TrkA, IC50=2 nM). Blocks TrkA-mediated γH2AX production and apoptosis in TrkA-overexpressing cells. Purity: ≥98% (TLC); mixture of E and Z isomers. Solubility: Soluble in DMSO (>25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0746