Learn More
Apexbio Technology LLC GW 583340 dihydrochloride 10mg
SDP
Catalog No. 5000088843
Encompass_Preferred
GW 583340 dihydrochloride (CAS 1173023-85-2) is a small molecule inhibitor targeting the tyrosine kinase activity of both EGFR and ErbB2 exhibiting potent inhibitory effects with IC50 values of 0 01 M and 0 014 M respectively In addition to suppressing these receptor-mediated signaling pathways GW 583340 dihydrochloride has been shown to inhibit ABC transporters including ABCG2 and ABCB1 thereby reversing transporter-associated multidrug resistance This compound is utilized in tumor biology research to investigate the modulation of oncogenic signaling mechanisms underlying drug resistance and to explore potential therapeutic strategies
Catalog No. 50-000-88843 Supplier Apexbio Technology LLC Supplier No. B698910
GW 583340 dihydrochloride (CAS 1173023-85-2) is a small molecule inhibitor targeting the tyrosine kinase activity of both EGFR and ErbB2 exhibiting potent inhibitory effects with IC50 values of 0 01 M and 0 014 M respectively In addition to suppressing these receptor-mediated signaling pathways GW 583340 dihydrochloride has been shown to inhibit ABC transporters including ABCG2 and ABCB1 thereby reversing transporter-associated multidrug resistance This compound is utilized in tumor biology research to investigate the modulation of oncogenic signaling mechanisms underlying drug resistance and to explore potential therapeutic strategies