Learn More
Abcam GW 590735, PPARalpha agonist, 25MG
Catalog No. 502705718
Encompass_Preferred
MW 478.5 Da, Purity >98%. Potent, selective PPARα agonist (EC₅₀ = 4 nM). Increases HDL cholesterol, decreases LDL and VLDL cholesterol. Reduces triglycerides. Shows antidyslipidemic effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5718 Supplier Abcam Supplier No. AB14444325MG
MW 478.5 Da, Purity >98%. Potent, selective PPARα agonist (EC₅₀ = 4 nM). Increases HDL cholesterol, decreases LDL and VLDL cholesterol. Reduces triglycerides. Shows antidyslipidemic effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement