Learn More
Selleck Chemical LLC GW 610-E0469-5MG
SDP
Catalog No. 503137551
Encompass_Preferred
GW 610 (NSC 721648) an antitumor agent shows potent and selective anticancer activity against lung colon and breast cancer cell lines
Catalog No. 50-313-7551 Supplier Selleck Chemical LLC Supplier No. E04695MG
GW 610 (NSC 721648) an antitumor agent shows potent and selective anticancer activity against lung colon and breast cancer cell lines