Learn More
Cayman Chemical GW 610 Neurochemicals
SDP
Catalog No. 502889973
Encompass_Preferred
An antitumor benzothiazole that shows growth-inhibitory activity against several cancer cell lines
Catalog No. 50-288-9973 Supplier Cayman Chemical Supplier No. 100083131 MG
An antitumor benzothiazole that shows growth-inhibitory activity against several cancer cell lines