Learn More
Cayman Chemical GW 610 Neurochemicals
SDP
Supplier:  Cayman Chemical 1000831350 MG
Encompass_Preferred
An antitumor benzothiazole that shows growth-inhibitory activity against several cancer cell lines
Catalog No. 50-289-1031