Learn More
Medchemexpress LLC HY-11083 10mg Medchemexpress, GW-803430 CAS:515141-51-2 Purity:>98%
SDP
Catalog No. 501968806
Encompass_Preferred
Medchemexpress, HY-11083 10mg GW-803430 CAS:515141-51-2 GW-803430 (GW-3430) is a potent and selective melanin-concentrating hormone receptor 1 (MCH R1) antagonist with a pIC50 of 9.3. GW-803430 is orally active in an animal model of obesity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8806 Supplier Medchemexpress LLC Supplier No. HY1108310MG
Medchemexpress, HY-11083 10mg GW-803430 CAS:515141-51-2 GW-803430 (GW-3430) is a potent and selective melanin-concentrating hormone receptor 1 (MCH R1) antagonist with a pIC50 of 9.3. GW-803430 is orally active in an animal model of obesity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.