Learn More
Medchemexpress LLC GW-870086 | 827319-43-7 | 99.6% | 559.64 | C31H39F2NO6 | 5 MG
SDP
Catalog No. 5000263764
Encompass_Preferred
GW-870086 is a synthetic glucocorticoid receptor agonist used in research to probe anti-inflammatory signaling. It inhibits NF-κB activity in A549 cells with a reported pIC50 of 10.1 and is supplied with high purity for in vitro pharmacology and cell-based assays.
  • Potent glucocorticoid receptor agonist active in cell-based NF-κB assays.
  • Reported pIC50 of 10.1 in A549 NF-κB assay.
  • High purity (99.59%) suitable for in vitro pharmacology.
  • Available as powder in multiple mg scales and as a 10 mM solution in DMSO for convenience.
  • Stable storage recommendations provided for powder and solvent solutions.
  • Molecular formula C31H39F2NO6; molecular weight 559.64.
Catalog No. 50-002-63764 Supplier Medchemexpress LLC Supplier No. HY1036625MG
GW-870086 is a synthetic glucocorticoid receptor agonist used in research to probe anti-inflammatory signaling. It inhibits NF-κB activity in A549 cells with a reported pIC50 of 10.1 and is supplied with high purity for in vitro pharmacology and cell-based assays.
  • Potent glucocorticoid receptor agonist active in cell-based NF-κB assays.
  • Reported pIC50 of 10.1 in A549 NF-κB assay.
  • High purity (99.59%) suitable for in vitro pharmacology.
  • Available as powder in multiple mg scales and as a 10 mM solution in DMSO for convenience.
  • Stable storage recommendations provided for powder and solvent solutions.
  • Molecular formula C31H39F2NO6; molecular weight 559.64.