Learn More
Selleck Chemical LLC GW0742-10mg
SDP
Supplier:  Selleck Chemical LLC S802010MG
Encompass_Preferred
GW0742 is a potent and highly selective PPARβ/δ agonist, with IC50 of 1 nM, with 1000-fold selectivity over hPPARα and hPPARγ.
Catalog No. 50-238-9997