Learn More
Apexbio Technology LLC GW0742, 10mg. Cas: 317318-84-6 MFCD: MFCD07369423. PPARδ/β agonist,potent and selective
SDP
Supplier:  Apexbio Technology LLC A430710
Encompass_Preferred
GW0742 is a synthetic, potent and selective PPAR-/ agonist. PPARs are ligand-dependent transcription factors which are involved in many physiological processes, such as inflammation and energy homeostasis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4914