Learn More
Apexbio Technology LLC GW0742 317318-84-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062575
Encompass_Preferred
GW0742 is a synthetic selective agonist of peroxisome proliferator-activated receptor / (PPAR / ) a ligand-dependent transcription factor involved in regulating lipid metabolism inflammation and energy homeostasis Activation of PPAR / modulates downstream gene transcription influencing physiological processes including inflammatory responses and neuronal protection Experimental studies demonstrate GW0742 s role in attenuating inflammation-mediated tissue injury such as ischemia/reperfusion-induced intestinal damage as well as mitigating radiation-induced neuroinflammation and cognitive impairment in animal models Thus GW0742 serves as a research tool to study PPAR / specific pathways in inflammatory diseases metabolic processes and neuroprotective mechanisms
Catalog No. 50-000-62575 Supplier Apexbio Technology LLC Supplier No. A43075.1
GW0742 is a synthetic selective agonist of peroxisome proliferator-activated receptor / (PPAR / ) a ligand-dependent transcription factor involved in regulating lipid metabolism inflammation and energy homeostasis Activation of PPAR / modulates downstream gene transcription influencing physiological processes including inflammatory responses and neuronal protection Experimental studies demonstrate GW0742 s role in attenuating inflammation-mediated tissue injury such as ischemia/reperfusion-induced intestinal damage as well as mitigating radiation-induced neuroinflammation and cognitive impairment in animal models Thus GW0742 serves as a research tool to study PPAR / specific pathways in inflammatory diseases metabolic processes and neuroprotective mechanisms