Learn More
Medchemexpress LLC HY-13928 5mg Medchemexpress, GW0742 CAS:317318-84-6 Purity:>98%
SDP
Catalog No. 502081532
Encompass_Preferred
Medchemexpress, HY-13928 5mg GW0742 CAS:317318-84-6 GW0742 is a potent PPARβ and PPARδ agonist, with an IC50 of 1 nM for human PPARδ in binding assay, and EC50s of 1 nM, 1.1 μM and 2 μM for human PPARδ, PPARα, and PPARγ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1532 Supplier Medchemexpress LLC Supplier No. HY139285MG
Medchemexpress, HY-13928 5mg GW0742 CAS:317318-84-6 GW0742 is a potent PPARβ and PPARδ agonist, with an IC50 of 1 nM for human PPARδ in binding assay, and EC50s of 1 nM, 1.1 μM and 2 μM for human PPARδ, PPARα, and PPARγ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.