Learn More
AdipoGen GW1929 (25 mg)
SDP
Supplier:  AdipoGen AGCR10116M025
Encompass_Preferred
Chemical. CAS: 196808-24-9. Formula: C30H29N3O4. MW: 495.6. Potent and subtype-selective (>1'000-fold) PPARgamma agonist. Angiogenesis inhibitor. Apoptosis inhibitor. Anti-inflammatory. Anti-hyperglycemic and anti-hyperlipidemic agent. Antidiabetic. The glucose-lowering effect in rats is 100-fold more potent than that of troglitazone.
Catalog No. 50-205-3844