Learn More
Enzo Life Sciences GW1929 (5mg). CAS: 196808-24-9
SDP
Supplier:  Enzo Life Sciences ALX420029M005
Encompass_Preferred
Potent (<10nM) and subtype-selective (>1'000-fold) PPARγ agonist which does not contain a thiazolidinedione moiety. The glucose-lowering effect of GW1929 in rats is 100-fold more potent than that of troglitazone (BML-GR210). Alternative name: N-(2-Benzoylphenyl)-L-tyrosine PPARgamma Agonist, (2S)-((2-Benzoylphenyl)amino-3-[4-[2-(methylpyridin-2-ylamino)ethoxy]phenyl)propionic acid. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9105