Learn More
AdipoGen GW1929
SDP
Catalog No. 502562364
Encompass_Preferred
Chemical. CAS 196808-24-9. Formula C30H29N3O4. MW 495.6. Potent and subtype-selective >1000-fold PPARgamma agonist. Angiogenesis inhibitor. Apoptosis inhibitor. Anti-inflammatory. Anti-hyperglycemic and anti-hyperlipidemic agent. Antidiabetic. The glucose-lowering effect in rats is 100-fold more potent than that of troglitazone.
Catalog No. 50-256-2364 Supplier AdipoGen Supplier No. AGCR10116M001
Chemical. CAS 196808-24-9. Formula C30H29N3O4. MW 495.6. Potent and subtype-selective >1000-fold PPARgamma agonist. Angiogenesis inhibitor. Apoptosis inhibitor. Anti-inflammatory. Anti-hyperglycemic and anti-hyperlipidemic agent. Antidiabetic. The glucose-lowering effect in rats is 100-fold more potent than that of troglitazone.