Learn More
Medchemexpress LLC HY-10917 10mg Medchemexpress, GW2580 CAS:870483-87-7 Purity:>98%
SDP
Catalog No. 501968689
Encompass_Preferred
Medchemexpress, HY-10917 10mg GW2580 CAS:870483-87-7 GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase which completely inhibits human cFMS kinase in vitro at 0.06 μM. GW2580 acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8689 Supplier Medchemexpress LLC Supplier No. HY1091710MG
Medchemexpress, HY-10917 10mg GW2580 CAS:870483-87-7 GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase which completely inhibits human cFMS kinase in vitro at 0.06 μM. GW2580 acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.