Learn More
Medchemexpress LLC HY-10917 1g , GW2580 CAS:870483-87-7 Purity:98%
SDP
Catalog No. 501492303
Encompass_Preferred
Medchemexpress, HY-10917 1g GW2580 CAS:870483-87-7 Formula:C20H22N4O3 IC50: 30 nM (c-FMS) Purity:98% GW2580 is a selective CSF-1R inhibitor for  c-FMS  with  IC 50  of 30 nM, and shows 150- to 500-fold selective compared to b-Raf, CDK4, c-KIT, c-SRC, EGFR, ERBB2/4, ERK2, FLT-3, GSK3, ITK, JAK2, etc. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2303 Supplier Medchemexpress LLC Supplier No. HY109171G
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-10917 1g GW2580 CAS:870483-87-7 Formula:C20H22N4O3 IC50: 30 nM (c-FMS) Purity:98% GW2580 is a selective CSF-1R inhibitor for  c-FMS  with  IC 50  of 30 nM, and shows 150- to 500-fold selective compared to b-Raf, CDK4, c-KIT, c-SRC, EGFR, ERBB2/4, ERK2, FLT-3, GSK3, ITK, JAK2, etc. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.