Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GW2580-d6 | 2733770-48-2 | 98.4% | 372.45 | 1 ML
SDP

Catalog No. 5000344614
Encompass_Preferred

GW2580-d6 is a deuterium-labeled version of GW2580, an orally bioavailable and selective inhibitor of c-Fms kinase. It completely inhibits human cFMS kinase in vitro at a concentration of 0.06 μM. This compound acts as a competitive inhibitor of ATP binding to the cFMS kinase, thereby inhibiting colony-stimulating-factor-1 signaling. Intended for research use only, it appears as a white to off-white solid.

  • Stable isotope: Contains deuterium labeling, useful as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 98.36% purity ensures reliable experimental results.
  • Selective inhibition: Potently inhibits c-Fms kinase, impacting colony-stimulating-factor-1 signaling.
  • Orally bioavailable: The unlabeled compound, GW2580, is orally bioavailable.

Catalog No. 50-003-44614 Supplier Medchemexpress LLC Supplier No. HY10917S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GW2580-d6 is a deuterium-labeled version of GW2580, an orally bioavailable and selective inhibitor of c-Fms kinase. It completely inhibits human cFMS kinase in vitro at a concentration of 0.06 μM. This compound acts as a competitive inhibitor of ATP binding to the cFMS kinase, thereby inhibiting colony-stimulating-factor-1 signaling. Intended for research use only, it appears as a white to off-white solid.

  • Stable isotope: Contains deuterium labeling, useful as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 98.36% purity ensures reliable experimental results.
  • Selective inhibition: Potently inhibits c-Fms kinase, impacting colony-stimulating-factor-1 signaling.
  • Orally bioavailable: The unlabeled compound, GW2580, is orally bioavailable.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.