Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GW2580-d6 | 2733770-48-2 | 98.4% | 372.45 g/mol | C20H16D6N4O3 | 5 MG
SDP

Catalog No. 5000263489
Encompass_Preferred

GW2580-d6 is the deuterium-labeled analogue of GW2580, a selective inhibitor of the colony-stimulating factor 1 receptor (CSF1R). Supplied as a white to off-white solid, this isotopically labeled standard is intended for use as an analytical reference, tracer, or internal standard in quantitative and pharmacokinetic studies.

  • Deuterium-labeled compound for use as an isotopic standard and tracer.
  • Selective CSF1R (c-Fms) inhibitor scaffold useful in kinase research.
  • Purity ~98.4% suitable for analytical applications.
  • Molecular weight 372.45 g/mol; formula includes six deuterium atoms.
  • Soluble in DMSO (~100 mg/mL); sonication and warming aid dissolution.
  • Powder stable at -20°C for long-term storage; store solutions frozen.

Catalog No. 50-002-63489 Supplier Medchemexpress LLC Supplier No. HY10917S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

GW2580-d6 is the deuterium-labeled analogue of GW2580, a selective inhibitor of the colony-stimulating factor 1 receptor (CSF1R). Supplied as a white to off-white solid, this isotopically labeled standard is intended for use as an analytical reference, tracer, or internal standard in quantitative and pharmacokinetic studies.

  • Deuterium-labeled compound for use as an isotopic standard and tracer.
  • Selective CSF1R (c-Fms) inhibitor scaffold useful in kinase research.
  • Purity ~98.4% suitable for analytical applications.
  • Molecular weight 372.45 g/mol; formula includes six deuterium atoms.
  • Soluble in DMSO (~100 mg/mL); sonication and warming aid dissolution.
  • Powder stable at -20°C for long-term storage; store solutions frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.