Learn More
Abcam GW2580 (DMSO solution), cFMS RTK inhibitor, 5MG
Supplier:  Abcam AB1466245MG
Encompass_Preferred
MW 366.4 Da, Purity >99%. Selective cFMS (colony stimulating factor 1) RTK inhibitor. Inhibits monocyte growth in vitro (IC₅₀ = 0.2 μM) and LPS-induced TNF-α production in vivo. Inhibits macrophage infiltration of the CNS. Orally active. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4545