Learn More
Medchemexpress LLC HY-114189 10mg Medchemexpress, GW284543 CAS:790186-68-4 Purity:>98%
SDP
Catalog No. 501969303
Encompass_Preferred
Medchemexpress, HY-114189 10mg GW284543 CAS:790186-68-4 GW284543 (UNC10225170) is a selective MEK5 inhibitor. GW284543 (UNC10225170) reduces pERK5, and decreases endogenous MYC protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9303 Supplier Medchemexpress LLC Supplier No. HY11418910MG
Medchemexpress, HY-114189 10mg GW284543 CAS:790186-68-4 GW284543 (UNC10225170) is a selective MEK5 inhibitor. GW284543 (UNC10225170) reduces pERK5, and decreases endogenous MYC protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.