Learn More
Medchemexpress LLC HY-15891 5mg Medchemexpress, GW311616 CAS:198062-54-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158915MG
Encompass_Preferred
Medchemexpress, HY-15891 5mg GW311616 CAS:198062-54-3 GW-311616 is a potent, orally bioavailable, long duration and selective human neutrophil elastase (HNE) inhibitor with IC50 value of 22 nM and Ki value of 0.31 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1819