Learn More
Apexbio Technology LLC GW311616 hydrochloride 197890-44-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088985
Encompass_Preferred
GW311616 hydrochloride (CAS 197890-44-1) is a selective cell-permeable inhibitor of human neutrophil elastase (HNE) exhibiting an IC50 of 22 nM and a Ki of 0 13 nM in enzymatic assays It demonstrates pronounced selectivity with IC50 values exceeding 100 M for trypsin cathepsin G and plasmin and greater than 3 M for chymotrypsin and tissue plasminogen activator In human whole blood assays GW311616 hydrochloride achieves an IC50 of 0 67 M The compound is orally bioavailable and displays prolonged activity in dog blood samples indicating a sustained inhibitory effect on neutrophil elastase GW311616 hydrochloride is valuable for investigating HNE s role in inflammation and related pathologies
Catalog No. 50-000-88985 Supplier Apexbio Technology LLC Supplier No. B46765.1
GW311616 hydrochloride (CAS 197890-44-1) is a selective cell-permeable inhibitor of human neutrophil elastase (HNE) exhibiting an IC50 of 22 nM and a Ki of 0 13 nM in enzymatic assays It demonstrates pronounced selectivity with IC50 values exceeding 100 M for trypsin cathepsin G and plasmin and greater than 3 M for chymotrypsin and tissue plasminogen activator In human whole blood assays GW311616 hydrochloride achieves an IC50 of 0 67 M The compound is orally bioavailable and displays prolonged activity in dog blood samples indicating a sustained inhibitory effect on neutrophil elastase GW311616 hydrochloride is valuable for investigating HNE s role in inflammation and related pathologies