Learn More
Medchemexpress LLC GW3965 50mg | 405911-09-3 | 50MG
SDP
Catalog No. 5000244260
Encompass_Preferred
GW3965 is a potent selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXR and hLXR respectively[1 [2 [3
Catalog No. 50-002-44260 Supplier Medchemexpress LLC Supplier No. HY1062750MG
GW3965 is a potent selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXR and hLXR respectively[1 [2 [3