Learn More
Apexbio Technology LLC GW3965 HCl 405911-17-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061712
Encompass_Preferred
GW3965 HCl is a selective non-steroidal agonist targeting liver X receptors (LXR and LXR ) nuclear transcription factors involved in cholesterol and lipid metabolism Activation of LXRs modulates cholesterol homeostasis by enhancing cholesterol efflux from macrophages stimulating hepatic bile acid synthesis and reducing intestinal cholesterol absorption GW3965 recruits steroid receptor coactivator-1 (SRC-1) to human LXRR in vitro and functions as a full agonist on cellular assays for both LXR and LXR receptors In animal studies oral administration elevated plasma HDL cholesterol levels and increased expression of the cholesterol transporter ABCA1 in macrophages GW3965 is useful in research investigating cholesterol metabolism atherosclerosis and vascular responses mediated by Angiotensin II signaling
Catalog No. 50-000-61712 Supplier Apexbio Technology LLC Supplier No. A84445.1
GW3965 HCl is a selective non-steroidal agonist targeting liver X receptors (LXR and LXR ) nuclear transcription factors involved in cholesterol and lipid metabolism Activation of LXRs modulates cholesterol homeostasis by enhancing cholesterol efflux from macrophages stimulating hepatic bile acid synthesis and reducing intestinal cholesterol absorption GW3965 recruits steroid receptor coactivator-1 (SRC-1) to human LXRR in vitro and functions as a full agonist on cellular assays for both LXR and LXR receptors In animal studies oral administration elevated plasma HDL cholesterol levels and increased expression of the cholesterol transporter ABCA1 in macrophages GW3965 is useful in research investigating cholesterol metabolism atherosclerosis and vascular responses mediated by Angiotensin II signaling