Learn More
Medchemexpress LLC HY-10627A 50mg , GW3965 (hydrochloride) CAS:405911-17-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10627A50MG
Encompass_Preferred
HY-10627A 50mg GW3965 (hydrochloride) CAS: 405911-17-3 GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9307