Learn More
Selleck Chemical LLC GW3965 HCl-5mg
SDP
Supplier:  Selleck Chemical LLC S26305MG
Encompass_Preferred
GW3965 HCl is a potent, selective LXR agonist for hLXRα and hLXRβ with EC50 of 190 and 30 nM in cell-free assays, respectively.
Catalog No. 50-238-9630