Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GW3965 hydrochloride | 405911-17-3 | MFCD08276920 | 100.0% | 618.51 | C33H32Cl2F3NO3 | 1 ML
SDP

Catalog No. 5000243766
Encompass_Preferred

GW3965 hydrochloride is the hydrochloride salt of GW3965, a potent and selective liver X receptor (LXR) agonist used in biochemical and cell-based research. It activates human LXRα and LXRβ with reported EC50 values of 190 nM and 30 nM, respectively.

  • Potent, selective LXR agonist with EC50s of 190 nM (hLXRα) and 30 nM (hLXRβ).
  • Suitable for in vitro and in vivo studies involving LXR activation.
  • Available as concentrated solution in DMSO and as solid for flexible dosing.
  • High purity supporting reproducible biological activity.
  • Soluble in DMSO and practically insoluble in water.
  • Storage recommendations provided for both solid and solution to preserve stability.

Catalog No. 50-002-43766 Supplier Medchemexpress LLC Supplier No. HY10627A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GW3965 hydrochloride is the hydrochloride salt of GW3965, a potent and selective liver X receptor (LXR) agonist used in biochemical and cell-based research. It activates human LXRα and LXRβ with reported EC50 values of 190 nM and 30 nM, respectively.

  • Potent, selective LXR agonist with EC50s of 190 nM (hLXRα) and 30 nM (hLXRβ).
  • Suitable for in vitro and in vivo studies involving LXR activation.
  • Available as concentrated solution in DMSO and as solid for flexible dosing.
  • High purity supporting reproducible biological activity.
  • Soluble in DMSO and practically insoluble in water.
  • Storage recommendations provided for both solid and solution to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.