Learn More
Medchemexpress LLC GW405833 hydrochloride | 1202865-22-2 | 99.59% | 483.82 | 1 ML
SDP
Catalog No. 5000343485
Encompass_Preferred
GW405833 (L768242) hydrochloride is a potent, selective cannabinoid receptor 2 (CB2) agonist. It has EC50 and Ki values of 0.65 nM and 3.92 nM for CB2, and EC50 and Ki values of 16.1 μM and 4772 nM for CB1. It also acts as a non-competitive CB1 antagonist, exerting its analgesic effect through a CB1 receptor dependent mechanism. It can significantly inhibit the production of cAMP stimulated by Forskolin and inhibits glycolysis by down-regulating HIF-1α, thereby alleviating acute liver failure (ALF).
  • Potent, selective cannabinoid receptor 2 (CB2) agonist
  • EC50 and Ki values for CB2: 0.65 nM and 3.92 nM respectively
  • EC50 and Ki values for CB1: 16.1 μM and 4772 nM respectively
  • Non-competitive CB1 antagonist
  • Inhibits cAMP production stimulated by Forskolin
  • Inhibits glycolysis by down-regulating HIF-1α
Catalog No. 50-003-43485 Supplier Medchemexpress LLC Supplier No. HY110036A50MG
GW405833 (L768242) hydrochloride is a potent, selective cannabinoid receptor 2 (CB2) agonist. It has EC50 and Ki values of 0.65 nM and 3.92 nM for CB2, and EC50 and Ki values of 16.1 μM and 4772 nM for CB1. It also acts as a non-competitive CB1 antagonist, exerting its analgesic effect through a CB1 receptor dependent mechanism. It can significantly inhibit the production of cAMP stimulated by Forskolin and inhibits glycolysis by down-regulating HIF-1α, thereby alleviating acute liver failure (ALF).
  • Potent, selective cannabinoid receptor 2 (CB2) agonist
  • EC50 and Ki values for CB2: 0.65 nM and 3.92 nM respectively
  • EC50 and Ki values for CB1: 16.1 μM and 4772 nM respectively
  • Non-competitive CB1 antagonist
  • Inhibits cAMP production stimulated by Forskolin
  • Inhibits glycolysis by down-regulating HIF-1α