Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GW405833 hydrochloride | 1202865-22-2 | MFCD09832650 | >98.0% | 483.82 g/mol | C23H25Cl3N2O3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY110036A5MG

Encompass_Preferred

GW405833 hydrochloride is the hydrochloride salt of a potent and selective cannabinoid receptor 2 (CB2) agonist used for pharmacology research. It is typically supplied as a white to off-white solid in milligram-scale quantities for in vitro and in vivo studies. Reported identifiers include CAS 1202865-22-2, molecular formula C23H25Cl3N2O3, and molecular weight ≈483.82 g/mol. Typical purity is ≥98% (HPLC).

  • Selective CB2 receptor agonist for receptor pharmacology studies.
  • Supplied as a solid for precise dosing in assays.
  • High purity (≥98.0%) for reliable experimental results.
  • Molecular weight 483.82 g/mol and formula C23H25Cl3N2O3.
  • Available in milligram-scale quantities for preclinical work.
  • Soluble in DMSO for in vitro assay preparation.

Catalog No. 50-002-63863


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.