Learn More
Medchemexpress LLC GW406108X 50mg | 1644443-92-4 | 400.21 | C20H11Cl2NO4 | 50 MG
SDP
Catalog No. 5000229019
Encompass_Preferred
GW406108X (GW108X) is a small-molecule research inhibitor of Kif15 and ULK1 used in cellular and biochemical studies of mitosis and autophagy. It is supplied as a high-purity research chemical (CAS 1644443-92-4) with molecular formula C20H11Cl2NO4 and molecular weight 400.21.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Reported activity against Kif15 and ULK1 for mechanistic studies.
  • Known molecular weight and formula to support compound characterization.
  • Supplied as a solid with good solubility in DMSO for stock preparation.
  • Available in small pack sizes appropriate for research-scale experiments.
Catalog No. 50-002-29019 Supplier Medchemexpress LLC Supplier No. HY11557050MG
GW406108X (GW108X) is a small-molecule research inhibitor of Kif15 and ULK1 used in cellular and biochemical studies of mitosis and autophagy. It is supplied as a high-purity research chemical (CAS 1644443-92-4) with molecular formula C20H11Cl2NO4 and molecular weight 400.21.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Reported activity against Kif15 and ULK1 for mechanistic studies.
  • Known molecular weight and formula to support compound characterization.
  • Supplied as a solid with good solubility in DMSO for stock preparation.
  • Available in small pack sizes appropriate for research-scale experiments.