Learn More
Selleck Chemical LLC GW441756 10mg 504433-23-2
SDP
Supplier:  Selleck Chemical LLC S289110MG
Encompass_Preferred
GW441756 is a potent, selective inhibitor of TrkA with IC50 of 2 nM, with very little activity to c-Raf1 and CDK2. GW441756 produces a relevant increase of caspase-3 that leads to apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0585504